Tetrasodium Pyrophosphate Decahydrate
Synonym(s):
Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, Tetrasodium pyrophosphate
Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing