Skip to Content
Merck
Polymyxin B solution 1 mg/mL in H2O, analytical standard

Polymyxin B Sulfate

Polymyxin B solution 1 mg/mL in H2O, analytical standard
Synonym(s):
Polymyxin B sulfate
CAS Number:
EC Number:

Products

Product Category
Brand
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique