Skip to Content
Merck
Salicylic acid phyproof® Reference Substance

Salicylic acid

Salicylic acid phyproof® Reference Substance
Synonym(s):
2-Hydroxybenzoic acid
Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique