Cyclosporine
Synonym(s):
Cyclosporin A, Antibiotic S 7481F1, Cyclosporine
Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing