Skip to Content
Merck
GTP >90% (HPLC), crystals, pkg of 250 mg

GTP

GTP >90% (HPLC), crystals, pkg of 250 mg
Synonym(s):
Guanosine 5′-triphosphate sodium salt hydrate, 5′-GTP-Na2
Empirical Formula (Hill Notation):
C10H16N5O14P3 · xNa+ · yH2O
CAS Number:
Molecular Weight:
523.18 (anhydrous free acid basis)

Products

Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing