![Doxazosin mesilate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/277/623/e3edbcb4-29a5-4a84-8a7b-8e3d9e2278a1/640/e3edbcb4-29a5-4a84-8a7b-8e3d9e2278a1.png)
Doxazosin mesilate
![Doxazosin mesilate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/277/623/e3edbcb4-29a5-4a84-8a7b-8e3d9e2278a1/640/e3edbcb4-29a5-4a84-8a7b-8e3d9e2278a1.png)
Synonym(s):
Doxazosin mesylate, 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[4-(1,4-benzodioxan-2-yl)carpiperazin-1-yl)]-6,7-dimethoxyquinazoline mesylate, Doxazosin methanesulfonate
Empirical Formula (Hill Notation):
C23H25N5O5 · CH3SO3H
CAS Number:
Molecular Weight:
547.58
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing