Skip to Content
Merck
BIOshell A160 Peptide CN, 2.7 μm HPLC Column 2.7 μm particle size, L × I.D. 10 cm × 3 mm

BIOshell A160 Peptide CN, 2.7 μm HPLC Column

BIOshell A160 Peptide CN, 2.7 μm HPLC Column 2.7 μm particle size, L × I.D. 10 cm × 3 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing