Skip to Content
Merck
Everolimus ≥95% (HPLC)

Everolimus

Everolimus ≥95% (HPLC)
Synonym(s):
Everolimus, 42-O-(2-Hydroxyethyl)rapamycin
Empirical Formula (Hill Notation):
C53H83NO14
CAS Number:
Molecular Weight:
958.22

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing