Urea
Synonym(s):
Carbamide, Carbonyldiamide, Urea solution, Urea
Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Products
Product Category
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade
Technique
Showing 1-20 of 27
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2