Skip to Content
Merck
(−)-Riboflavin from Eremothecium ashbyii, ≥98%

(−)-Riboflavin

(−)-Riboflavin from Eremothecium ashbyii, ≥98%
Synonym(s):
Lactoflavin, Vitamin B2, Vitamin G, Riboflavine, Riboflavin
Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Technique