N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide
Synonym(s):
MTBSTFA, N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide, MTBSTFA + 1% TBDMSCl, Silylating mixture VI, N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide
Linear Formula:
CF3CON(CH3)Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
241.33
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Showing 1-5 of 5