Merck
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C (1 min), 10 °C/min to 325 °C (5 min)
inj. temp.
300 °C
detector
FID, 365 °C
MSD interface
300 °C
scan range
m/z 45-450
carrier gas
helium, 0.9 mL/min., constant
injection
1 μL, splitless (1 min)
liner
3.8 mm I.D., straight
sample
Each analyte at 10 μg/mL in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany