Merck
GC Analysis of Class 3 Residual Solvents on OVI-G43
GC Analysis of Class 3 Residual Solvents on OVI-G43 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
OVI-G43 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 3.00 μm
CONDITIONS
column
OVI-G43, 30 m x 0.53 mm I.D., 3.0 μm with 5 m intermediate polarity guard (25396)
oven
35 °C (15 min), 5 °C/min to 200 °C
inj. temp.
225 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec @ 35 °C
injection
1 μL 33:1 split
liner
single taper
sample
27 analytes, each at 3000 μg/mL in methanol