Merck
  • Defective Kernel 39 encodes a PPR protein required for seed development in maize.
Defective Kernel 39 encodes a PPR protein required for seed development in maize.
Journal of integrative plant biology (2017-10-06)
Xiaojie Li, Wei Gu, Silong Sun, Zongliang Chen, Jing Chen, Weibin Song, Haiming Zhao, Jinsheng Lai
ABSTRACT
RNA editing is a posttranscriptional process that is important in mitochondria and plastids of higher plants. All RNA editing-specific trans-factors reported so far belong to PLS-class of pentatricopeptide repeat (PPR) proteins. Here, we report the map-based cloning and molecular characterization of a defective kernel mutant dek39 in maize. Loss of Dek39 function leads to delayed embryogenesis and endosperm development, reduced kernel size, and seedling lethality. Dek39 encodes an E sub-class PPR protein that targets to both mitochondria and chloroplasts, and is involved in RNA editing in mitochondrial NADH dehydrogenase3 (nad3) at nad3-247 and nad3-275. C-to-U editing of nad3-275 is not conserved and even lost in Arabidopsis, consistent with the idea that no close DEK39 homologs are present in Arabidopsis. However, the amino acids generated by editing nad3-247 and nad3-275 are highly conserved in many other plant species, and the reductions of editing at these two sites decrease the activity of mitochondria NADH dehydrogenase complex I, indicating that the alteration of amino acid sequence is necessary for Nad3 function. Our results indicate that Dek39 encodes an E sub-class PPR protein that is involved in RNA editing of multiple sites and is necessary for seed development of maize.
Sigma-Aldrich
HistoChoice® Clearing Agent, alternative to toluene and xylene