Merck
  • Light-Enhanced Antibacterial Activity of Graphene Oxide, Mainly via Accelerated Electron Transfer.
Light-Enhanced Antibacterial Activity of Graphene Oxide, Mainly via Accelerated Electron Transfer.
Environmental science & technology (2017-08-05)
Yu Chong, Cuicui Ge, Ge Fang, Renfei Wu, He Zhang, Zhifang Chai, Chunying Chen, Jun-Jie Yin
ABSTRACT
Before graphene derivatives can be exploited as next-generation antimicrobials, we must understand their behavior under environmental conditions. Here, we demonstrate how exposure to simulated sunlight significantly enhances the antibacterial activity of graphene oxide (GO) and reveal the underlying mechanism. Our measurements of reactive oxygen species (ROS) showed that only singlet oxygen (