Merck
  • Novel genetic aberrations in breast phyllodes tumours: comparison between prognostically distinct groups.
Novel genetic aberrations in breast phyllodes tumours: comparison between prognostically distinct groups.
Breast cancer research and treatment (2014-05-17)
Wai Jin Tan, Johnathan C Lai, Aye Aye Thike, Jeffrey Chun Tatt Lim, Sie Yong Tan, Valerie Cui Yun Koh, Tse Hui Lim, Boon Huat Bay, Min-Han Tan, Puay Hoon Tan
ABSTRACT
Phyllodes tumours of the breast are uncommon fibroepithelial neoplasms which pose management challenges due to difficulties in accurate prediction of clinical behaviour, as histological assessment has its limitations. Molecular studies have improved the understanding of these rare tumours but such findings are scant. We aimed to investigate genetic aberrations in phyllodes tumours stratified according to clinical behaviour, to identify potential genes contributing to disease progression. Twenty phyllodes tumours were separated into prognostically distinct categories depending on whether they had recurred/metastasized within the follow-up period. DNA extracted from FFPE materials was subjected to Affymetrix OncoScan™ FFPE Express molecular inversion probe microarray platform for analysis of copy number changes and mutational status. Results were cross validated with Sanger sequencing, FISH and immunohistochemistry. A higher number of chromosomal aberrations were observed in cases which recurred/metastasized, with median events of 19 compared to 3.5 in cases which did not recur/metastasize. High-level amplification and homozygous deletions were detected exclusively in the former group. Regions of high-level amplification included MDM4 (1q32.1), RAF1 (3p25), EGFR (7p12) and PDZD2 (5p13.3). EGFR amplification was confirmed on FISH and accompanied by intense EGFR immunostaining. Regions of homozygous deletion included CDKN2A (9p21) and MACROD2 (20p12.1). Homozygous deletion of 9p21 which involved CDKN2A was accompanied by loss of protein expression. No mutations were identified in all samples. These findings provide insights into identifying target genes and pathways exploited by phyllodes tumours, which would aid future development of individualised therapy.