Merck
  • Adjuvant activities of novel cytokines, interleukin-23 (IL-23) and IL-27, for induction of hepatitis C virus-specific cytotoxic T lymphocytes in HLA-A*0201 transgenic mice.
Adjuvant activities of novel cytokines, interleukin-23 (IL-23) and IL-27, for induction of hepatitis C virus-specific cytotoxic T lymphocytes in HLA-A*0201 transgenic mice.
Journal of virology (2004-08-17)
Masanori Matsui, Osamu Moriya, Maria Laura Belladonna, Sadahiro Kamiya, François A Lemonnier, Takayuki Yoshimoto, Toshitaka Akatsuka
ABSTRACT
Searching the sequence databases has revealed two novel cytokines: interleukin-23 (IL-23) and IL-27. These cytokines are quite similar to, but clearly distinct from IL-12 in their structures and T-cell stimulatory fashions. In contrast to IL-12, however, little is known about the roles of IL-23 and IL-27 in the immune regulation. Previously, we evaluated the prime-boost immunization consisting of priming and the first boosting with the hepatitis C virus (HCV)-core expression plasmid, followed by a second boosting with recombinant adenovirus expressing HCV core for induction of HCV core-specific cytotoxic T lymphocytes (CTLs) in BALB/c mice. The present study demonstrates that HCV-specific CTL induction was greatly enhanced by coinoculation of an IL-12 expression plasmid in the prime-boost immunization, indicating the potent adjuvant activity of IL-12. We investigated whether similar adjuvant effects could be exerted by either IL-23 or IL-27 in a prime-boost immunization with HLA-A*0201 transgenic mice. Coadministration of either an IL-23 or an IL-27 expression plasmid, as well as an IL-12 expression plasmid, in a prime-boost immunization enhanced induction of HCV-specific CTLs and led to dramatic increases in the numbers of gamma interferon (IFN-gamma)-producing, HCV-specific CD8+ cells. Further, preinjections of IL-12, IL-23, or IL-27 expression plasmids before immunization resulted in great increases in the number of IFN-gamma-producing, HCV-specific CD8+ cells in response to immunization with recombinant adenovirus. These data revealed that both IL-23 and IL-27, as well as IL-12, are potent adjuvants for epitope-specific CTL induction. The two novel cytokines might offer new prophylactic and therapeutic strategies against infectious pathogens such as HCV.