Merck
Avasimibe Pfizer.
Current opinion in investigational drugs (London, England : 2000) (2002-12-25)
John R Burnett, Murray W Huff
ABSTRACT
Avasimibe (also known as CI-1011 or PD-148515) is a selective acyl-coenzyme A:cholesterol O-acyltransferase (ACAT) inhibitory lipid-regulating agent under development by Pfizer (formerly Parke-Davis) for the potential treatment of hyperlipidemia and atherosclerosis. The compound is currently undergoing phase III clinical trials [371470].
Sigma-Aldrich
Avasimibe, ≥98% (HPLC)