- Flavonol glycosides from Dryas octopetala.
Flavonol glycosides from Dryas octopetala.
Journal of natural products (1984-09-01)
O Servettaz, M L Colombo, M De Bernardi, E Uberti, G Vidari, P Vita-Finzi
PMID6512533
ABSTRACT
Six flavonol glycosides and ent-epicatechin were isolated from Dryas octopetala and their structures elucidated by chemical and spectroscopic methods. Two new flavonoids, 3-O-alpha-L-arabinofuranosyl-8-methoxyquercetin (2) and 3-O-beta-galactopyranosyl-8-methoxykaempferol (6), were identified along with 3-O-beta-D-galactopyranosylquercetin (hyperin) (3), 3-O-alpha-L-arabinofuranosylquercetin (avicularin) (5), 3-O-beta-L-arabinopyranosylquercetin (4), and 3-O-beta-D-galactopyranosyl-8-methoxyquercetin (1).