Merck
  • Inhibitory activities of three classes of acyclic nucleoside phosphonates against murine polyomavirus and primate simian virus 40 strains.
Inhibitory activities of three classes of acyclic nucleoside phosphonates against murine polyomavirus and primate simian virus 40 strains.
Antimicrobial agents and chemotherapy (2007-04-11)
Ilya Lebeau, Graciela Andrei, Marcela Krecmerová, Erik De Clercq, Antonin Holy, Robert Snoeck
ABSTRACT
Murine polyomavirus and simian virus 40 were used to evaluate the potencies of the compounds of three classes of acyclic nucleoside phosphonates: (i) the original HPMP (3-hydroxy-2-phosphonomethoxypropyl) and PME (2-phosphonomethoxyethyl) derivatives, (ii) the 6-[2-(phosphonomethoxy)alkoxy]-2,4-diaminopyrimidine (DAPy) derivatives, and (iii) a new class of HPMP derivatives containing a 5-azacytosine moiety. The last class showed the highest activities and selectivities against both polyomaviruses.
Sigma-Aldrich
2,4-Diaminopyrimidine, 98%