Merck
  • SENP1 participates in Irinotecan resistance in human colon cancer cells.
SENP1 participates in Irinotecan resistance in human colon cancer cells.
Journal of cellular biochemistry (2021-05-27)
Ming-Cheng Chen, Do Chi Nhan, Chiung-Hung Hsu, Tso-Fu Wang, Chi-Cheng Li, Tsung-Jung Ho, B Mahalakshmi, Mei-Chih Chen, Liang-Yo Yang, Chih-Yang Huang
ABSTRACT
Colorectal cancer is one of the most prevalent cancers in the world. Chemoresistance has always been a problem encountered in its treatment. It is known that SUMOylation may regulate protein stability and decomposition, and even affect the protein translocation and posttranslational modification in cells. Sentrin-specific protease 1 (SENP1) is involved in the maturation of SUMO protein, and on the other hand, plays a role in deSUMOylation, which dissociates the target protein from SUMO and prevents further degradation of the target protein. In this study, we established an Irinotecan (CPT-11) resistant human colon cancer LoVo strain (LoVoR-CPT-11 ) to investigate the role of SENP1 in the development of drug resistance in colorectal cancer. The abundant accumulation of SENP1 and HIF-1α proteins and the increase of SUMO pathway enzymes were observed in LoVoR-CPT-11 cells while the protein markers of proliferation, angiogenesis, and glycolysis were upregulated. Knockdown of SENP1 reduced the migration ability and trigged re-sensitivity of LoVoR-CPT-11 cells to CPT-11 treatment. The analysis of SENP1 and HIF-1α gene expressions from TCGA/GTEx datasets using the GEPIA web server showed a positive correlation between SENP1 and HIF-1α in colorectal cancer patients and the high expression of these two genes might predict a poor outcome clinically. In conclusion, SENP1 might play an important role in CPT-11 resistance in colorectal cancer. Targeting SENP1 to reduce the resistant property could be considered in prospective clinical studies.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
Anti-19S regulator non-ATPase subunit S5a/Rpn10 Antibody, clone S5a-18, clone S5a-18, Upstate®, from mouse