Merck
  • Subcritical Methanol Extraction of the Stone of Japanese Apricot Prunus mume Sieb. et Zucc.
Subcritical Methanol Extraction of the Stone of Japanese Apricot Prunus mume Sieb. et Zucc.
Biomolecules (2020-07-19)
Tomoaki Kawabata, Yoshie Tanaka, Asako Horinishi, Megumi Mori, Asao Hosoda, Nami Yamamoto, Takahiko Mitani
ABSTRACT
The pits of Japanese apricot, Prunus mume Sieb. et Zucc., which are composed of stones, husks, kernels, and seeds, are unused by-products of the processing industry in Japan. The processing of Japanese apricot fruits generates huge amounts of waste pits, which are disposed of in landfills or, to a lesser extent, burned to form charcoal. Mume stones mainly consist of cellulose, hemicellulose, and lignin. Herein, we attempted to solubilize the wood-like carapace (stone) encasing the pit by subcritical fluid extraction with the aim of extracting useful chemicals. The characteristics of the main phenolic constituents were elucidated by liquid chromatography-mass spectrometry (LC-MS) and nuclear magnetic resonance (NMR) analyses. The degrees of solubility for various treatments (190 °C; 3 h) were determined as follows: subcritical water (54.9%), subcritical 50% methanol (65.5%), subcritical 90% methanol (37.6%), subcritical methanol (23.6%), and subcritical isopropyl alcohol (14.4%). Syringaldehyde, sinapyl alcohol, coniferyl alcohol methyl ether, sinapyl alcohol methyl ether, 5-(hydroxymethyl)-2-furfural, and furfural were present in the subcritical 90% methanol extract. Coniferyl and sinapyl alcohols (monolignols) are source materials for the biosynthesis of lignin, and syringaldehyde occur in trace amounts in wood. Our current findings provide a solubilization method that allows the main phenolic constituents of the pits to be extracted under mild conditions. This technique for obtaining subcritical extracts shows great potential for further applications.
Sigma-Aldrich
Sinapyl alcohol, technical grade, 80%