- Nicotinic acetylcholine receptor subunit mRNA expression and channel function in medial habenula neurons.
Nicotinic acetylcholine receptor subunit mRNA expression and channel function in medial habenula neurons.
Relationships between nicotinic acetylcholine receptor (nAChR) channel function and nAChR subunit mRNA expression were explored in acutely isolated rat medial habenula (MHb) neurons using a combination of whole-cell recording and single cell RT-PCR techniques. Following amplification using subunit-specific primers, subunits could be categorized in one of three ways: (i) present in 95-100% cells: alpha3, alpha4, alpha5, beta2 and beta4; (ii) never present: alpha2; and (iii) sometimes present ( approximately 40% cells): alpha6, alpha7 and beta3. These data imply that alpha2 subunits do not participate in nAChRs on MHb cells, that alpha6, alpha7 and beta3 subunits are not necessary for functional channels but may contribute in some cells, and that nAChRs may require combinations of all or subsets of alpha3, alpha4, alpha5, beta2 and beta4 subunits. Little difference in the patterns of subunit expression between nicotine-sensitive and insensitive cells were revealed based on this qualitative analysis, implying that gene transcription per se may be an insufficient determinant of nAChR channel function. Normalization of nAChR subunit levels to the amount of actin mRNA, however, revealed that cells with functional channels were associated with high levels (>0.78 relative to actin; 11/12 cells) of all of the category (i) subunits: alpha3, alpha4, alpha5, beta2 and beta4. Conversely, one or more of these subunits was always low (<0.40 relative to actin) in all cells with no detectable response to nicotine. Thus the formation of functional nAChR channels on MHb cells may require critical levels of several subunit mRNAs.