Skip to Content
Merck
Glucosamine for system suitability European Pharmacopoeia (EP) Reference Standard

Glucosamine hydrochloride

Glucosamine for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
D-(+)-Glucosamine hydrochloride, 2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride
Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique