Skip to Content
KZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium chloride solution
Linear Formula:
MgCl
2
CAS Number:
7786-30-3
Molecular Weight:
95.21
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (5)
Color
Formula Weight
Manufacturer
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Application
Package Size
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
M1028
Magnesium chloride solution,
for molecular biology, 1.00 M±0.01 M
Expand
View Pricing
68475
Magnesium chloride solution,
BioUltra, for molecular biology, 2 M in H
2
O
Expand
View Pricing
63069
Magnesium chloride solution,
BioUltra, for molecular biology, ~1 M in H
2
O
Expand
View Pricing
M8787
Magnesium chloride solution,
PCR Reagent, 25 mM MgCI
2
solution for PCR
Expand
View Pricing
11699113001
Magnesium chloride solution,
pH 8.25-8.45 (25 °C), suitable for PCR, solution, pkg of 3 × 1 mL
Expand
View Pricing
00457
Magnesium chloride solution,
0.1 M
Expand
View Pricing
63036
Magnesium chloride solution,
BioUltra, for molecular biology, ~0.025 M in H
2
O
Expand
View Pricing