Propidium iodide solution
Synonym(s):
3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide, 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium, diiodide, Propidium Iodide Solution, Propidium iodide solution
Empirical Formula (Hill Notation):
C27H34I2N4
CAS Number:
Molecular Weight:
668.39
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Detection Method
Quality Segment
Application
Technique
Showing 1-4 of 4