
Lipopolysaccharides from Escherichia coli O111:B4

Synonym(s):
LPS
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Research Area
Quality Segment
Application
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing