Skip to Content
Merck
L-Asparagine tert-butyl ester hydrochloride ≥98.0% (TLC)

L-Asparagine tert-butyl ester hydrochloride

L-Asparagine tert-butyl ester hydrochloride ≥98.0% (TLC)
Linear Formula:
NH2COCH2CH(NH2)COOC(CH3)3 · HCl
CAS Number:
Molecular Weight:
224.69

Products

Brand
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing