![Gonyautoxin-2 and Gonyautoxin-3 solution 20 μg/g in acetic acid (nominal concentration), analytical standard](/deepweb/assets/sigmaaldrich/product/images/203/395/bfb09dc6-b505-456a-98e8-8c5683142185/640/bfb09dc6-b505-456a-98e8-8c5683142185.jpg)
Gonyautoxin-2 and Gonyautoxin-3 solution
![Gonyautoxin-2 and Gonyautoxin-3 solution 20 μg/g in acetic acid (nominal concentration), analytical standard](/deepweb/assets/sigmaaldrich/product/images/203/395/bfb09dc6-b505-456a-98e8-8c5683142185/640/bfb09dc6-b505-456a-98e8-8c5683142185.jpg)
Synonym(s):
GTX-2 - GTX-3 mixture
Empirical Formula (Hill Notation):
C10H17N7O8S
Molecular Weight:
395.35
Products
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing