Skip to Content
Merck
3-Amino-1,2,4-triazole ≥95% (TLC)

3-Amino-1,2,4-triazole

3-Amino-1,2,4-triazole ≥95% (TLC)
Synonym(s):
1,2,4-Triazol-3-amine, 3-AT, Amitrol, 3-Amino-1H-1,2,4-triazole
Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08

Products

Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing