Skip to Content
KZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4arm-PEG5K-NH2
Synonym(s):
4arm-PEG-NH2
Linear Formula:
C(CH
2
O(CH
2
CH
2
O)nCH
2
CH
2
NH
2
HCl)
4
Products
Brand
Sigma-Aldrich (1)
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
JKA7020
4arm-PEG5K-NH2,
HCl Salt, average M
n
5,000
Expand
View Pricing