Magnesium nitrate hexahydrate
Synonym(s):
Magnesium nitrate hexahydrate, Magnesium nitrate – nitric acid solution
Linear Formula:
Mg(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
256.41
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-8 of 8
Compare
Product Number
Description
Pricing