Skip to Content
Merck
Isopropyl β-D-1-thiogalactopyranoside

IPTG

Isopropyl β-D-1-thiogalactopyranoside
Synonym(s):
Isopropyl β-D-1-thiogalactopyranoside, IPTG, Isopropyl β-D-thiogalactoside, IPTG, Dioxane-Free, High Purity, OmniPur® IPTG, IPTG, Animal-Free, High Purity
Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Technique