Vancomycin hydrochloride from Streptomyces orientalis
Synonym(s):
Vancomycin hydrochloride, CSB Selective Supplement (Vancomycin)
Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · xHCl
CAS Number:
Molecular Weight:
1449.25 (free base basis)
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Research Area
Quality Segment
Application
Microorganism Suitability
Mode of Action
Sterility
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing