Merck
Scepter™ Cell Counter
Scepter™ 3.0 Handheld Automated Cell Counter
The Scepter™ 3.0 Cell Counter is the only portable, handheld automated cell counter currently available. While other automated counters consume bench space and rely on hand-filled loading chambers, the Scepter® 3.0 Cell Counter provides true automation using Coulter impedance counting. With its microfluidic sensor, the Scepter® 3.0 Cell Counter counts thousands of cells per sample, and does the work of assessment and calculation to deliver accurate, consistent cell counts in less than 30 seconds.
  • Large, easy-to-read display
  • Wireless data transmission to printer or lab workstation
  • USB port for data access & transfer
  • Validated in 88 cell lines and multiple applications
  • 2 sensor sizes: 40 µm and 60 µm
  • Results in less than 30 seconds
Already have a Scepter™ 2.0 Cell Counter? Learn how you can upgrade to the Scepter™ 3.0 Cell Counter.
Scepter™ Cell Counter Application Notes
Learn how Scepter™ Cell Counter has been used in a variety of applications, including somatic cell counting in dairy milk samples, human PBMC counting, and tracking ADSC and 3T3-L1 cell differentiation.