Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702246

Sigma-Aldrich

Anti-SARS-CoV-2 N Protein ,BSA and glycerol freeAntibody, clone 4Z6T2, Rabbit Monoclonal

Synonym(s):

Nucleoprotein, NP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

material

colorless

clone

4Z6T2, monoclonal

form

liquid

species reactivity

SARS coronavirus

concentration

1.9mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200

color

colorless

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

SARS coronavirus ... Nucleoprotein(1489678)

General description

Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M. Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication. May modulate transforming growth factor-beta signaling by binding host SMAD3.

Immunogen

PBS with 0.05% proclin300,pH7.3.
Recombinant fusion protein of SARS-CoV-2 Nucleoprotein.

Application

DB, WB, IF/ICC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service