Merck
HPLC Analysis of Thalidomide Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Thalidomide Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materials
분석 컬럼
제품 번호
설명
가격
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
표준 물질
제품 번호
설명
가격
(+)-Thalidomide
≥98% (HPLC), powder
(±)-Thalidomide
≥98%, powder
(−)-Thalidomide
>98%, solid
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 18 mM ammonium nitrate, pH 5.5; [B] acetonitrile; (65:35, A:B)
flow rate
1 mL/min
column temp.
22 °C
detector
UV, 240 nm
injection
20 μL