Merck
SGE autosampler syringes, fixed needle, for CTC/LEAP 10F-C/F-GT-0.63, Slim line, Gas tight, volume 10 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
SGE autosampler syringes, fixed needle, for CTC/LEAP
SGE autosampler syringes, fixed needle, for CTC/LEAP 10F-C/F-GT-0.63, Slim line, Gas tight, volume 10 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm