뒤로
Hellma
®
absorption cuvettes, standard cells, ultra Micro
Products
제품 카테고리
cuvettes (4)
Manufacturer
직경(mm)
높이(mm)
Volume (µL)
검출 방법
Technique
제품 필터
Showing 1-4 of 4
비교
제품 번호
설명
가격
Z600393
Hellma
®
absorption cuvettes, standard cells, ultra Micro,
High Performance Quartz Glass, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 50 μL
확장
가격 보기
Z600415
Hellma
®
absorption cuvettes, standard cells, ultra Micro,
High Performance Quartz Glass, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 50 μL
확장
가격 보기
Z627046
Hellma
®
absorption cuvettes, standard cells, ultra Micro,
High Performance Quartz Glass, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 5 μL
확장
가격 보기
Z627062
Hellma
®
absorption cuvettes, standard cells, ultra Micro,
High Performance Quartz Glass, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 5 μL