주요 문서
DWKKC92812G-2000
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with KIMCOTE® safety coating and glass stopper
glass flask, capacity 2,000 mL, class A
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121804
NACRES:
NB.43
material
glass flask
특징
class A
포장
case of 1 ea
제조업체/상표
Kimble® KC92812G-2000
용량
2,000 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
KimCote Class A Heavy Duty Wide Mouth Volumetric Flasks
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
특징 및 장점
  • Complies with ASTM Specification E288, Class A requirements
  • Manufactured from borosilicate glass 3.3
  • Autoclavable KimCote safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Wide mouth volumetric flasks feature neck dimensions that will easily accommodate pipets
  • Superior strength, durability and heavy uniform walls ensure safety in the lab
  • Calibrated to contain
  • Flat interior bottoms are ideal for stirring with standard stir bars
법적 정보
KIMBLE is a registered trademark of DWK Life Sciences
KIMCOTE is a registered trademark of DWK LIFE SCIENCES LLC
Kimax is a registered trademark of Gerresheimer Glass, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)