HPLC Analysis of 2,3-Dihydroxy-3-Phenylpropionic Acid Enantiomers on CHIROBIOTIC R
Materials
analytical column
CONDITIONS
column
CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 μm particles (13024AST)
mobile phase
[A] 0.1% ammonia, pH 4.1 with formic acid; [B] methanol; (50:50, A:B)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 258 nm
injection
10 μL
sample
2,3-dihydroxy-3-phenyl-propionic acid isomers:
(A) racemate A (4.85 and 6.95 min)
(B) racemate B (5.33 and 6.29 min)