Merck
  • Targeting of Arabidopsis KNL2 to Centromeres Depends on the Conserved CENPC-k Motif in Its C Terminus.
Targeting of Arabidopsis KNL2 to Centromeres Depends on the Conserved CENPC-k Motif in Its C Terminus.
The Plant cell (2017-01-08)
Michael Sandmann, Paul Talbert, Dmitri Demidov, Markus Kuhlmann, Twan Rutten, Udo Conrad, Inna Lermontova
ABSTRACT
KINETOCHORE NULL2 (KNL2) is involved in recognition of centromeres and in centromeric localization of the centromere-specific histone cenH3. Our study revealed a cenH3 nucleosome binding CENPC-k motif at the C terminus of