Merck
  • Deletion of the β-acetoacetyl synthase FabY in Pseudomonas aeruginosa induces hypoacylation of lipopolysaccharide and increases antimicrobial susceptibility.
Deletion of the β-acetoacetyl synthase FabY in Pseudomonas aeruginosa induces hypoacylation of lipopolysaccharide and increases antimicrobial susceptibility.
Antimicrobial agents and chemotherapy (2013-10-23)
David A Six, Yanqiu Yuan, Jennifer A Leeds, Timothy C Meredith
ABSTRACT
The β-acetoacetyl-acyl carrier protein synthase FabY is a key enzyme in the initiation of fatty acid biosynthesis in Pseudomonas aeruginosa. Deletion of fabY results in an increased susceptibility of P. aeruginosa in vitro to a number of antibiotics, including vancomycin and cephalosporins. Because antibiotic susceptibility can be influenced by changes in membrane lipid composition, we determined the total fatty acid profile of the ΔfabY mutant, which suggested alterations in the lipid A region of the lipopolysaccharide. The majority of lipid A species in the ΔfabY mutant lacked a single secondary lauroyl group, resulting in hypoacylated lipid A. Adding exogenous fatty acids to the growth media restored the wild-type antibiotic susceptibility profile and the wild-type lipid A fatty acid profile. We suggest that incorporation of hypoacylated lipid A species into the outer membrane contributes to the shift in the antibiotic susceptibility profile of the ΔfabY mutant.