Merck
  • Phospholipase D is a central regulator of collagen I-induced cytoskeletal rearrangement and podosome formation in megakaryocytes.
Phospholipase D is a central regulator of collagen I-induced cytoskeletal rearrangement and podosome formation in megakaryocytes.
Journal of thrombosis and haemostasis : JTH (2014-06-10)
S Stritt, I Thielmann, S Dütting, D Stegner, B Nieswandt
ABSTRACT
Blood platelets are small anucleated cell fragments generated from bone marrow megakaryocytes (MKs) by a cytoskeleton-driven process. Thereby, mature MKs form long cytoplasmic protrusions (pro-platelets), which extend into the sinusoids within the bone marrow and finally release platelets. Podosomes are F-actin rich matrix contacts that have been suggested to play an important role in cell migration, but also in pro-platelet formation by MKs. Phospholipase D (PLD) has been proposed to contribute to the regulation of actin dynamics through the local generation of phosphatidic acid but its role in platelet formation is unknown. We sought to investigate the significance of PLD in MK podosome formation and thrombocytopoiesis. Podosome formation, spreading and ultra-structure of PLD single- and double-deficient MKs were analyzed using confocal and transmission electron microscopy. Phospholipase D-deficient MKs displayed a highly altered ultra-structure in vivo and abnormal actin rearrangement, with almost abolished formation of podosomes upon spreading on collagen I in vitro. However, MK endomitosis and platelet production were not altered by PLD deficiency. Together, our findings point to a specific function of PLD in actin dynamics as well as podosome formation and size determination in MKs on a collagen I matrix. The normal platelet number in PLD-deficient mice, however, suggests the existence of compensatory mechanisms in vivo that overcome the defective podosome formation observed in vitro.