Merck
  • The feasibility of enhanced soil washing of p-nitrochlorobenzene (pNCB) with SDBS/Tween80 mixed surfactants.
The feasibility of enhanced soil washing of p-nitrochlorobenzene (pNCB) with SDBS/Tween80 mixed surfactants.
Journal of hazardous materials (2009-06-23)
Huiqin Guo, Zhenyu Liu, Shaogui Yang, Cheng Sun
ABSTRACT
The present study investigated the feasibility of using two mixed surfactants, anionic surfactant sodium dodecylbenzenesulfonate (SDBS) and nonionic surfactant polysorbate 80 (Tween80), for the remediation of p-nitrochlorobenzene (pNCB) contaminated soil. The water solubility, the apparent soil-water distribution constant (K(d)(*)) and the desorption ratio of pNCB, as well as the sorption of surfactants by the soil were significantly affected by the dosage of surfactants and the mass ratio of SDBS/Tween80. Because of the formation of mixed micelles, the presence of SDBS showed more effective than individual Tween80 for increasing the water solubility, decreasing the K(d)(*) and enhancing the desorption ratio of pNCB, as well as inhibiting the sorption of surfactants by the soil. Low dosage of surfactants (Tween80 < 2000 mg L(-1)) increased the K(d)(*) value and inhibited the desorption of pNCB from soil. However, relative high concentration of Tween80 had positive effect on the decrease of the K(d)(*) value and increase of pNCB desorption. In addition, among the tested surfactant systems, mixed SDBS/Tween80 with a 1:1 mass ratio exhibited the highest pNCB desorption. The results indicated that it is feasible to use mixed SDBS/Tween80 surfactants for the remediation of pNCB contaminated soil.
Sigma-Aldrich
1-Chloro-4-nitrobenzene, 99%