- B-Cell Epitope Mapping Using a Library of Overlapping Synthetic Peptides in an Enzyme-Linked Immunosorbent Assay.
B-Cell Epitope Mapping Using a Library of Overlapping Synthetic Peptides in an Enzyme-Linked Immunosorbent Assay.
Methods in molecular biology (Clifton, N.J.) (2018-05-02)
Thiru Vanniasinkam, Mary D Barton, Tongted Phumoonna Das, Michael W Heuzenroeder
PMID29714015
ABSTRACT
This chapter describes a strategy for mapping linear B-cell epitopes of proteins using synthetic biotinylated peptides in an ELISA.A set of overlapping peptides were designed based upon a known amino acid sequence of the target protein, VapA (Virulence-associated Protein A) of the bacterium Rhodococcus equi, an important pulmonary pathogen in foals. The peptides synthesized as biotinylated peptides were coated directly onto micro titer plates which had been pre-coated with NeutrAvidin™ and used to screen sera from foals confirmed to have R. equi disease. A linear B-cell epitope was identified which corresponded to a 20 mer sequence of the VapA protein.