Merck
  • Identification and validation of cryptochrome inhibitors that modulate the molecular circadian clock.
Identification and validation of cryptochrome inhibitors that modulate the molecular circadian clock.
ACS chemical biology (2014-01-07)
Sung Kook Chun, Jaebong Jang, Sooyoung Chung, Hwayoung Yun, Nam-Jung Kim, Jong-Wha Jung, Gi Hoon Son, Young-Ger Suh, Kyungjin Kim
ABSTRACT
Circadian rhythms, biological oscillations with a period of about 24 h, are maintained by a genetically determined innate time-keeping system called the molecular circadian clockwork. Despite the physiological and clinical importance of the circadian clock, the development of small molecule modulators that directly target the core clock machinery has only been recently initiated. In the present study, we aimed to identify novel small molecule modulators influencing the molecular feedback loop of the circadian clock by applying our two-step cell-based screening strategy based on E-box-mediated transcriptional activity to test more than 1000 drug-like compounds. A derivative of 2-ethoxypropanoic acid designated as compound 15 was selected as the most promising candidate in terms of both efficacy and potency. We then performed pull-down assays with the biotinylated compound and find out that both cryptochrome (CRY)1 and 2 (CRY1/2), key negative components of the mammalian circadian clock, as molecular targets of compound 15. In accordance with the binding property, compound 15 enhanced E-box-mediated transcription in a CRY1/2-dependent manner, and more importantly, it attenuated the circadian oscillation of Per2-Luc and Bmal1-dLuc activities in cultured fibroblasts, indicating that compound 15 can functionally inhibit the effects of CRY1/2 in the molecular circadian clockwork. In conclusion, the present study describes the first novel chemical inhibitor of CRY1/2 that inhibits the repressive function of CRY1/2, thereby activating CLOCK-BMAL1-evoked E-box-mediated transcription. Further optimizations and subsequent functional studies of this compound may lead to development of efficient therapeutic strategies for a variety of physiological and metabolic disorders with circadian natures.
Sigma-Aldrich
5-(Biotinamido)pentylamine TFA Salt, ≥95%