2-Methyl-1-propanol
Synonym(s):
Isobutanol, Isobutyl alcohol
Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Products
Shipping
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing