Skip to Content
Merck
Testosterone for system suitability A CRS, European Pharmacopoeia (EP) Reference Standard

Testosterone

Testosterone for system suitability A CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Testosterone, 17β-Hydroxy-3-oxo-4-androstene, 17β-Hydroxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one, trans-Testosterone
Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade