Skip to Content
Merck
Lithium aluminum hydride solution 0.5 M in 2-methoxyethyl ether

Lithium aluminum hydride solution

Lithium aluminum hydride solution 0.5 M in 2-methoxyethyl ether
Synonym(s):
LAH, Lithium alanate, Lithium tetrahydroaluminate
Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95

Products

Shipping
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Reagent Type